1,1'-Diethyl-2,2'-cyanine iodide, 99%, Thermo Scientific Chemicals
1,1'-Diethyl-2,2'-cyanine iodide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,1'-Diethyl-2,2'-cyanine iodide, 99%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
500 mg
Packaging:
Glass bottle
Catalog number 407255000
Price (USD)/ Each
104.00
-
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
104.00
Add to cart
1,1'-Diethyl-2,2'-cyanine iodide, 99%, Thermo Scientific Chemicals
Catalog number407255000
Price (USD)/ Each
104.00
-
Add to cart
Chemical Identifiers
CAS977-96-8
IUPAC Name1-ethyl-2-[(1-ethyl-1,2-dihydroquinolin-2-ylidene)methyl]quinolin-1-ium iodide
Molecular FormulaC23H23IN2
InChI KeyGMYRVMSXMHEDTL-UHFFFAOYSA-M
SMILES[I-].CCN1C(=CC2=CC=C3C=CC=CC3=[N+]2CC)C=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)Golden to dark red
UV spectrophotometry>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only