Thermo Scientific Chemicals

Diethyl iminodiacetate, 98%, Thermo Scientific Chemicals

Catalog number: 407321000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Diethyl iminodiacetate, 98%, Thermo Scientific Chemicals

Catalog number: 407321000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 407321000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6290-05-7
IUPAC Name
ethyl 2-[(2-ethoxy-2-oxoethyl)amino]acetate
Molecular Formula
C8H15NO4
InChI Key
LJDNMOCAQVXVKY-UHFFFAOYSA-N
SMILES
CCOC(=O)CNCC(=O)OCC
Appearance (Form)
Liquid
Refractive index
1.4350 to 1.4380 (20°C, 589 nm)
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to yellow
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text