Diethyl oxalacetate sodium salt, 95%, pract., Thermo Scientific Chemicals
Diethyl oxalacetate sodium salt, 95%, pract., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl oxalacetate sodium salt, 95%, pract., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 407375000
Price (USD)/ Each
140.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
165.00 
Save 24.35 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
140.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
165.00 
Save 24.35 (15%)
Add to cart
Diethyl oxalacetate sodium salt, 95%, pract., Thermo Scientific Chemicals
Catalog number407375000
Price (USD)/ Each
140.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
165.00 
Save 24.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namesodium 1,4-diethoxy-1,3,4-trioxobutan-2-ide
Molecular FormulaC8H11NaO5
InChI KeyJPTKZRPOIUYFTM-UHFFFAOYSA-N
SMILES[Na+].CCOC(=O)[CH-]C(=O)C(=O)OCC
Molecular Weight (g/mol)210.16
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)Beige to light orange
Infrared spectrumConforms
Titration with HClO4>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only