Thermo Scientific Chemicals

N,N-Diethyl-p-phenylenediamine, 98%, Thermo Scientific Chemicals

Catalog number: 407391000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

N,N-Diethyl-p-phenylenediamine, 98%, Thermo Scientific Chemicals

Catalog number: 407391000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 407391000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
93-05-0
IUPAC Name
N1,N1-diethylbenzene-1,4-diamine
Molecular Formula
C10H16N2
InChI Key
QNGVNLMMEQUVQK-UHFFFAOYSA-N
SMILES
CCN(CC)C1=CC=C(N)C=C1
GC
>=97.5 %
Appearance (Color)
Clear yellow-brown to dark brown to black
Infrared spectrum
Conforms
Appearance (Form)
Liquid or low melting solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text