D-(+)-Sucrose, crystals, NF, Multi-Compendial, GMP, J.T.Baker™
D-(+)-Sucrose, crystals, NF, Multi-Compendial, GMP, J.T.Baker™
Avantor™
D-(+)-Sucrose, crystals, NF, Multi-Compendial, GMP, J.T.Baker™
This product is GMP Manufactured and meets JP, BP, EP and NF Specifications.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4074-2050 kg
4074-01500 g
4074-052.5 kg
4074-0712 kg
4074-1990 kg
Catalog number 4074-20
Price (EUR)
-
Request A Quote
Quantity:
50 kg
Ask our AI about this Product
This product is GMP Manufactured and meets JP, BP, EP and NF Specifications.
Specifications
Quantity50 kg
Molecular Weight (g/mol)342.3
GradeMulti-Compendial/NF
Physical FormCrystalline Powder
Unit Size50 kg
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link