1x1 image pixel for data collection
Digitonin
Digitonin
Digitonin
Thermo Scientific Chemicals

Digitonin

CAS: 11024-24-1 | C56H92O29 | 1229.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
5 g
Packaging:
Glass bottle
Catalog number 407560050
Price (USD)
1,565.65
Online Exclusive
1,739.00
Save 173.35 (10%)
Each
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
1,565.65
Online Exclusive
1,739.00
Save 173.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS11024-24-1
SpecificationsSpecification SheetSpecification Sheet
Residue after ignition=<0.3 %
Appearance (Color)White to yellow
Appearance of solution(2.5% in warm EtOH) clear colorless to faint yellow
Infrared spectrumConforms
Specific optical rotation-45° to -51° (20°C, 589 nm) (c=10, 75% acetic acid)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link