Dihexylamine, 99+%, Thermo Scientific Chemicals
Dihexylamine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dihexylamine, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number 407595000
Price (USD)/ Each
175.00
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
175.00
Add to cart
Dihexylamine, 99+%, Thermo Scientific Chemicals
Catalog number407595000
Price (USD)/ Each
175.00
-
Add to cart
Chemical Identifiers
CAS143-16-8
IUPAC Namedihexylamine
Molecular FormulaC12H27N
InChI KeyPXSXRABJBXYMFT-UHFFFAOYSA-N
SMILESCCCCCCNCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Refractive index1.4320 to 1.4340 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only