7,12-Dimethylbenz[a]anthracene, 98%
7,12-Dimethylbenz[a]anthracene, 98%
7,12-Dimethylbenz[a]anthracene, 98%
7,12-Dimethylbenz[a]anthracene, 98%
Thermo Scientific Chemicals

7,12-Dimethylbenz[a]anthracene, 98%

CAS: 57-97-6 | C20H16 | 256.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
5 g
Packaging:
Glass bottle
Catalog number 408181000
Price (USD)
54.65
Special offer
Online exclusive
Ends: 26-Sep-2025
67.20
Save 12.55 (19%)
Each
In stock
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
54.65
Special offer
Online exclusive
Ends: 26-Sep-2025
67.20
Save 12.55 (19%)
Each
Add to cart
Chemical Identifiers
CAS57-97-6
IUPAC Name7,12-dimethyltetraphene
Molecular FormulaC20H16
InChI KeyARSRBNBHOADGJU-UHFFFAOYSA-N
SMILESCC1=C2C=CC3=CC=CC=C3C2=C(C)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to green
Infrared spectrumConforms
HPLC>=97.5 %
Melting point120°C to 124°C
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link