Dimethyldioctadecylammonium bromide, 99+%, Thermo Scientific Chemicals
Dimethyldioctadecylammonium bromide, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyldioctadecylammonium bromide, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 408260050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3700-67-2
IUPAC Namedimethyldioctadecylazanium bromide
Molecular FormulaC38H80BrN
InChI KeyPSLWZOIUBRXAQW-UHFFFAOYSA-M
SMILES[Br-].CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Melting point>=150°C clear point
TLC>=99.0 %
Appearance (Form)Powder and/or chunks
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only