N,N-Dimethyldodecylamine, 95%, pract., Thermo Scientific Chemicals
N,N-Dimethyldodecylamine, 95%, pract., Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Dimethyldodecylamine, 95%, pract., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Glass bottle
Catalog number 408290010
Price (USD)/ Each
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS112-18-5
IUPAC Namedodecyldimethylamine
Molecular FormulaC14H31N
InChI KeyYWFWDNVOPHGWMX-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Titration>=94 % (Tertiary amine)
GC>=94 % (Dodecyl)
Appearance (Color)Clear to slightly hazy
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only