Thermo Scientific Chemicals

Dimethylglyoxime, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 408330025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Dimethylglyoxime, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 408330025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 408330025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
95-45-4
IUPAC Name
(E)-N-[(3E)-3-(hydroxyimino)butan-2-ylidene]hydroxylamine
Molecular Formula
C4H8N2O2
InChI Key
JGUQDUKBUKFFRO-GGWOSOGESA-N
SMILES
C\C(=N/O)\C(\C)=N\O
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder
Suitability
(for Ni determination) Passes test
Infrared spectrum
Conforms
Gravimetry
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text