Thermo Scientific Chemicals

Dimethylglyoxime, disodium salt octahydrate, 99%, Thermo Scientific Chemicals

Catalog number: 408341000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Dimethylglyoxime, disodium salt octahydrate, 99%, Thermo Scientific Chemicals

Catalog number: 408341000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 408341000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75006-64-3
IUPAC Name
disodium oxido[3-(oxidoimino)butan-2-ylidene]amine octahydrate
Molecular Formula
C4H22N2Na2O10
InChI Key
IJJDRGUPRATWQX-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.O.[Na+].[Na+].CC(=N[O-])C(C)=N[O-]
Appearance (Color)
White to light beige
Infrared spectrum
Conforms
Water
43 to 48 % (K.F.)
Sensitivity
(Test for Ni) Passes
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text