Thermo Scientific Chemicals

N,N-Dimethylguanidine sulfate, 98%, Thermo Scientific Chemicals

Catalog number: 408351000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

N,N-Dimethylguanidine sulfate, 98%, Thermo Scientific Chemicals

Catalog number: 408351000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 408351000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
598-65-2
IUPAC Name
N,N-dimethylguanidine; sulfuric acid
Molecular Formula
C3H11N3O4S
InChI Key
ACPFLXJEPKOPGF-UHFFFAOYSA-N
SMILES
OS(O)(=O)=O.CN(C)C(N)=N
Appearance (Form)
Powder
Titration with HClO4
>=97.5 %
Infrared spectrum
Conforms
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text