1,4-Dinitrobenzene, 98%, Thermo Scientific Chemicals
1,4-Dinitrobenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Dinitrobenzene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 408650050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name1,4-dinitrobenzene
Molecular FormulaC6H4N2O4
InChI KeyFYFDQJRXFWGIBS-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(C=C1)[N+]([O-])=O
Molecular Weight (g/mol)168.11
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to brown
Appearance (Form)Powder or crystals and/or chunks
Melting point171°C to 176°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only