Thermo Scientific Chemicals

3,4-Dinitrobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 408700050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

3,4-Dinitrobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 408700050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 408700050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
528-45-0
IUPAC Name
3,4-dinitrobenzoic acid
Molecular Formula
C7H4N2O6
InChI Key
OMVRRHJJQILIJX-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(C(=C1)[N+]([O-])=O)[N+]([O-])=O
Appearance (Color)
Yellow
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
Melting point
163°C to 166°C
Titration with NaOH
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text