Thermo Scientific Chemicals

Methyldiphenylphosphine, 99%, Thermo Scientific Chemicals

Catalog number: 408970050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyldiphenylphosphine, 99%, Thermo Scientific Chemicals

Catalog number: 408970050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 408970050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1486-28-8
IUPAC Name
methyldiphenylphosphane
Molecular Formula
C13H13P
InChI Key
UJNZOIKQAUQOCN-UHFFFAOYSA-N
SMILES
CP(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.6230 to 1.6260 (20°C, 589 nm)
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text