Methyldiphenylphosphine, 99%, Thermo Scientific Chemicals
Methyldiphenylphosphine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyldiphenylphosphine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 408971000
Price (USD)/ Each
620.00
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
620.00
Add to cart
Methyldiphenylphosphine, 99%, Thermo Scientific Chemicals
Catalog number408971000
Price (USD)/ Each
620.00
-
Add to cart
Chemical Identifiers
CAS1486-28-8
IUPAC Namemethyldiphenylphosphane
Molecular FormulaC13H13P
InChI KeyUJNZOIKQAUQOCN-UHFFFAOYSA-N
SMILESCP(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.6230 to 1.6260 (20°C, 589 nm)
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only