Diphenylphosphoryl azide, 98%, Thermo Scientific Chemicals
Diphenylphosphoryl azide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diphenylphosphoryl azide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 409041000
Price (USD)/ Each
207.65
Online exclusive
230.00 
Save 22.35 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
207.65
Online exclusive
230.00 
Save 22.35 (10%)
Add to cart
Diphenylphosphoryl azide, 98%, Thermo Scientific Chemicals
Catalog number409041000
Price (USD)/ Each
207.65
Online exclusive
230.00 
Save 22.35 (10%)
-
Add to cart
Chemical Identifiers
CAS26386-88-9
IUPAC Name{[azido(phenoxy)phosphoryl]oxy}benzene
Molecular FormulaC12H10N3O3P
InChI KeySORGEQQSQGNZFI-UHFFFAOYSA-N
SMILES[N-]=[N+]=NP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5500 to 1.5540 (20°C, 589 nm)
Appearance (Color)Clear yellow to amber
GC>=97.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, For Azidation/Diazo transfer
RUO – Research Use Only