Dipropyl phthalate, 99+%, Thermo Scientific Chemicals
Dipropyl phthalate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dipropyl phthalate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 409160250
Price (USD)/ Each
84.65
Online exclusive
93.90 
Save 9.25 (10%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
84.65
Online exclusive
93.90 
Save 9.25 (10%)
Add to cart
Dipropyl phthalate, 99+%, Thermo Scientific Chemicals
Catalog number409160250
Price (USD)/ Each
84.65
Online exclusive
93.90 
Save 9.25 (10%)
-
Add to cart
Chemical Identifiers
CAS131-16-8
IUPAC Name1,2-dipropyl benzene-1,2-dicarboxylate
Molecular FormulaC14H18O4
InChI KeyMQHNKCZKNAJROC-UHFFFAOYSA-N
SMILESCCCOC(=O)C1=CC=CC=C1C(=O)OCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=99.0 %
Refractive index1.4960 to 1.4980 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only