DL-1,4-Dithiothreitol, 98%, Pure
DL-1,4-Dithiothreitol, 98%, Pure
Thermo Scientific Chemicals

DL-1,4-Dithiothreitol, 98%, Pure

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 409190010
Price (USD)/ Each
47.65
Online exclusive
52.70 
Save 5.05 (10%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
47.65
Online exclusive
52.70 
Save 5.05 (10%)
Add to cart
DL-1,4-Dithiothreitol, 98%, Pure
Catalog number409190010
Price (USD)/ Each
47.65
Online exclusive
52.70 
Save 5.05 (10%)
-
Add to cart
Chemical Identifiers
CAS3483-12-3
IUPAC Name(2R,3R)-1,4-disulfanylbutane-2,3-diol
Molecular FormulaC4H10O2S2
InChI KeyVHJLVAABSRFDPM-IMJSIDKUSA-N
SMILESO[C@@H](CS)[C@@H](O)CS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting point38°C to 43°C
Infrared spectrumConforms
Appearance (Form)Powder or beads
Titration Iodimetric97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only