Thermo Scientific Chemicals

Dodecyltrimethylammonium bromide, 99%, pure, Thermo Scientific Chemicals

Catalog number: 409310250
25 g, Each
Thermo Scientific Chemicals

Dodecyltrimethylammonium bromide, 99%, pure, Thermo Scientific Chemicals

Catalog number: 409310250
25 g, Each
Quantity
Catalog number: 409310250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1119-94-4
IUPAC Name
dodecyltrimethylazanium bromide
Molecular Formula
C15H34BrN
InChI Key
XJWSAJYUBXQQDR-UHFFFAOYSA-M
SMILES
[Br-].CCCCCCCCCCCC[N+](C)(C)C
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
Color of solution
=<100 APHA (10% aq. soln.)
Appearance (Form)
Powder
Titration Argentometric
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text