Thermo Scientific Chemicals

Dodecyltrimethylammonium chloride, 98%, Thermo Scientific Chemicals

Catalog number: 409320250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Dodecyltrimethylammonium chloride, 98%, Thermo Scientific Chemicals

Catalog number: 409320250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 409320250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
112-00-5
IUPAC Name
dodecyltrimethylazanium chloride
Molecular Formula
C15H34ClN
InChI Key
DDXLVDQZPFLQMZ-UHFFFAOYSA-M
SMILES
[Cl-].CCCCCCCCCCCC[N+](C)(C)C
Appearance (Form)
Powder and/or chunks
Appearance (Color)
White to light yellow
Titration Argentometric
>=97.5 %
Solubility
(10 % in water) Clear
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text