Eosin Y, pure, certified
Eosin Y, pure, certified
Eosin Y, pure, certified
Eosin Y, pure, certified
Thermo Scientific Chemicals

Eosin Y, pure, certified

CAS: 17372-87-1 | C20H12Br4Na2O8 | 745.904 g/mol
Catalog NumberQuantity
40943025025 g
Catalog number 409430250
Price (JPY)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS17372-87-1
IUPAC Namedisodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular FormulaC20H6Br4Na2O5
InChI KeySEACYXSIPDVVMV-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Br)C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(Br)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red to brown
Dye content>=88 %
BSC CertificationPasses
Infrared spectrumConforms
UV Lambda max515 to 518 nm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Share catalog number, name or link