Thermo Scientific Chemicals

Erythrosin B, pure, certified

Catalog number: 409450250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Erythrosin B, pure, certified

Catalog number: 409450250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 409450250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16423-68-0
IUPAC Name
disodium 2-(2,4,5,7-tetraiodo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular Formula
C20H6I4Na2O5
InChI Key
IINNWAYUJNWZRM-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]C(=O)C1=C(C=CC=C1)C1=C2C=C(I)C(=O)C(I)=C2OC2=C(I)C([O-])=C(I)C=C12
Appearance (Color)
Red to brown
Infrared spectrum
Conforms
UV Lambda max
524 to 527 nm
Appearance (Form)
Powder
BSC Certification
Passes

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text