Thermo Scientific Chemicals

4'-Ethoxyacetophenone, 99%, Thermo Scientific Chemicals

Catalog number: 409595000
500 g, Each
Thermo Scientific Chemicals

4'-Ethoxyacetophenone, 99%, Thermo Scientific Chemicals

Catalog number: 409595000
500 g, Each
Quantity
Catalog number: 409595000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1676-63-7
IUPAC Name
1-(4-ethoxyphenyl)ethan-1-one
Molecular Formula
C10H12O2
InChI Key
YJFNFQHMQJCPRG-UHFFFAOYSA-N
SMILES
CCOC1=CC=C(C=C1)C(C)=O
Melting point
35°C to 38°C
Appearance (Color)
White to yellow
Infrared spectrum
Conforms
GC
>=98.5 %
Appearance (Form)
Crystals and/or chunks or low melting solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text