Thermo Scientific Chemicals

Ethoxyamine hydrochloride, 99+%, Thermo Scientific Chemicals

Catalog number: 409610100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethoxyamine hydrochloride, 99+%, Thermo Scientific Chemicals

Catalog number: 409610100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 409610100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3332-29-4
IUPAC Name
hydrogen O-ethylhydroxylamine chloride
Molecular Formula
C2H8ClNO
InChI Key
NUXCOKIYARRTDC-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CCON
Appearance (Form)
Crystalline powder or crystals and/or chunks
Appearance (Color)
White to light yellow
Titration Argentometric
97.5 to 102.5 %
Melting point
128°C to 138°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text