4-(4-Ethoxyphenylazo)-m-phenylenediamine, mixture of mono and dihydrochloride, 98%, Thermo Scientific Chemicals
4-(4-Ethoxyphenylazo)-m-phenylenediamine, mixture of mono and dihydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(4-Ethoxyphenylazo)-m-phenylenediamine, mixture of mono and dihydrochloride, 98%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
10 g
Packaging:
Glass bottle
Catalog number 409680100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS74367-87-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark brown to black
Infrared spectrumConforms
Appearance (Form)Powder
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only