Ethyl 2-bromo-3-methylbutyrate, 95%, Thermo Scientific Chemicals
Ethyl 2-bromo-3-methylbutyrate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-bromo-3-methylbutyrate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
View all
Glass Bottle
Catalog number 409790500
Price (USD)/ Each
204.00
-
Add to cart
Quantity:
50 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
204.00
Add to cart
Ethyl 2-bromo-3-methylbutyrate, 95%, Thermo Scientific Chemicals
Catalog number409790500
Price (USD)/ Each
204.00
-
Add to cart
Chemical Identifiers
CAS609-12-1
IUPAC Nameethyl 2-bromo-3-methylbutanoate
Molecular FormulaC7H13BrO2
InChI KeyWNFUWONOILPKNX-UHFFFAOYNA-N
SMILESCCOC(=O)C(Br)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4480 to 1.4510 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only