2-Ethylbutyryl chloride, 99%, Thermo Scientific Chemicals
2-Ethylbutyryl chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Ethylbutyryl chloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 409821000
Price (USD)/ Each
219.00
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
219.00
Add to cart
2-Ethylbutyryl chloride, 99%, Thermo Scientific Chemicals
Catalog number409821000
Price (USD)/ Each
219.00
-
Add to cart
Chemical Identifiers
CAS2736-40-5
IUPAC Name2-ethylbutanoyl chloride
Molecular FormulaC6H11ClO
InChI KeySMUKODJVMQOSAB-UHFFFAOYSA-N
SMILESCCC(CC)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only