Thermo Scientific Chemicals

Ethylenebis(oxyethylenenitrilo)tetraacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 409910250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Ethylenebis(oxyethylenenitrilo)tetraacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 409910250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 409910250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67-42-5
IUPAC Name
3,12-bis(carboxymethyl)-6,9-dioxa-3,12-diazatetradecanedioic acid
Molecular Formula
C14H24N2O10
InChI Key
DEFVIWRASFVYLL-UHFFFAOYSA-N
SMILES
OC(=O)CN(CCOCCOCCN(CC(O)=O)CC(O)=O)CC(O)=O
Infrared spectrum
Conforms
Titration excess NaOH
>=97.5 %
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Melting point
240°C to 244°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text