Thermo Scientific Chemicals

Ethylene dimethacrylate, 98%, stabilized, Thermo Scientific Chemicals

Catalog number: 409920051
5 L, Plastic drum, Each
Thermo Scientific Chemicals

Ethylene dimethacrylate, 98%, stabilized, Thermo Scientific Chemicals

Catalog number: 409920051
5 L, Plastic drum, Each
Quantity
Packaging
Catalog number: 409920051
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
97-90-5
IUPAC Name
2-[(2-methylprop-2-enoyl)oxy]ethyl 2-methylprop-2-enoate
Molecular Formula
C10H14O4
InChI Key
STVZJERGLQHEKB-UHFFFAOYSA-N
SMILES
CC(=C)C(=O)OCCOC(=O)C(C)=C
Infrared spectrum
Conforms
Refractive index
1.4530 to 1.4550 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text