Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, ACS reagent

Catalog number: 409975000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, ACS reagent

Catalog number: 409975000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 409975000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6381-92-6
IUPAC Name
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
Molecular Formula
C10H18N2Na2O10
InChI Key
OVBJJZOQPCKUOR-UHFFFAOYSA-L
SMILES
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Infrared spectrum
Conforms
pH
4.0 to 6.0 (5 % at 25°C)
Appearance (Color)
White to light yellow
Titration Complexometric
99.0 to 101.0 %
Insoluble matter
=<0.005 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text