Thermo Scientific Chemicals

Ethylene glycol, 95%, pure, Thermo Scientific Chemicals

Catalog number: 410010010
1 kg, Glass Bottle, Each
Thermo Scientific Chemicals

Ethylene glycol, 95%, pure, Thermo Scientific Chemicals

Catalog number: 410010010
1 kg, Glass Bottle, Each
Quantity
Packaging
Catalog number: 410010010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-21-1
IUPAC Name
ethane-1,2-diol
Molecular Formula
C2H6O2
InChI Key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES
OCCO
Color scale
=<20 APHA
GC
>=94 %
Appearance (Form)
Clear viscous liquid
Infrared spectrum
Conforms
Refractive index
1.4300 to 1.4330 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text