2-Ethylhexyl acetate, 99%, pure, Thermo Scientific Chemicals
2-Ethylhexyl acetate, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Ethylhexyl acetate, 99%, pure, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
500 g
Packaging:
Glass bottle
Catalog number 410105000
Price (USD)/ Each
46.70
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
46.70
Add to cart
2-Ethylhexyl acetate, 99%, pure, Thermo Scientific Chemicals
Catalog number410105000
Price (USD)/ Each
46.70
-
Add to cart
Chemical Identifiers
CAS103-09-3
IUPAC Name2-ethylhexyl acetate
Molecular FormulaC10H20O2
InChI KeyWOYWLLHHWAMFCB-UHFFFAOYNA-N
SMILESCCCCC(CC)COC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4190 to 1.4210 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only