Thermo Scientific Chemicals

Ethylidene diacetate, 99%, Thermo Scientific Chemicals

Catalog number: 410131000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethylidene diacetate, 99%, Thermo Scientific Chemicals

Catalog number: 410131000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 410131000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
542-10-9
IUPAC Name
1-(acetyloxy)ethyl acetate
Molecular Formula
C6H10O4
InChI Key
ACKALUBLCWJVNB-UHFFFAOYSA-N
SMILES
CC(OC(C)=O)OC(C)=O
Infrared spectrum
Conforms
Appearance (Form)
Liquid
Refractive index
1.4000 to 1.4020 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text