Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Choose Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks for use with a wide range of lab reagents and solutions due to their excellent chemical compatibility. Rugged, shatterproof construction and light weight compared with glass make these flasks a valuble element of lab safety programs.
Capacity (English) | 32 oz. |
Description | Flask Erlenmeyer PP 1000mL |
Capacity (Metric) | 1000 mL |
Material | PPCO |
No. per Case | 6 |
No. per Pack | 2 |
Autoclavable | Autoclavable |
Disposable | Reusable |
Graduated | No |
Mouth Style | Narrow |
Sterility | Non-sterile |
Bottom Shape | Plain |
Closure Size | Stopper No. 8 |
Unit Size | Case of 6 |
Capacity (English) | 4 oz. |
Description | Flask Erlenmeyer PP 125mL |
Capacity (Metric) | 125 mL |
Material | PPCO |
No. per Case | 24 |
No. per Pack | 12 |
Autoclavable | Autoclavable |
Disposable | Reusable |
Graduated | No |
Mouth Style | Narrow |
Sterility | Non-sterile |
Bottom Shape | Plain |
Closure Size | Stopper No. 3 |
Unit Size | Case of 24 |
Capacity (English) | 4 oz. |
Description | Flask Erlenmeyer PP 125mL |
Capacity (Metric) | 125 mL |
Material | PPCO |
No. per Case | 24 |
No. per Pack | 12 |
Autoclavable | Autoclavable |
Disposable | Reusable |
Graduated | No |
Mouth Style | Narrow |
Sterility | Non-sterile |
Bottom Shape | Plain |
Closure Size | Stopper No. 3 |
Unit Size | Pack of 12 |
Capacity (English) | 8 oz. |
Description | Flask Erlenmeyer PP 250mL |
Capacity (Metric) | 250 mL |
Material | PPCO |
No. per Case | 12 |
No. per Pack | 6 |
Autoclavable | Autoclavable |
Disposable | Reusable |
Graduated | No |
Mouth Style | Narrow |
Sterility | Non-sterile |
Bottom Shape | Plain |
Closure Size | Stopper No. 6 |
Unit Size | Case of 12 |
Capacity (English) | 8 oz. |
Description | Flask Erlenmeyer PP 250mL |
Capacity (Metric) | 250 mL |
Material | PPCO |
No. per Case | 12 |
No. per Pack | 6 |
Autoclavable | Autoclavable |
Disposable | Reusable |
Graduated | No |
Mouth Style | Narrow |
Sterility | Non-sterile |
Bottom Shape | Plain |
Closure Size | Stopper No. 6 |
Unit Size | Pack of 6 |
Capacity (English) | 32 oz. |
Description | Flask Erlenmeyer PP 1000mL |
Capacity (Metric) | 1000 mL |
Material | PPCO |
No. per Case | 6 |
No. per Pack | 2 |
Autoclavable | Autoclavable |
Disposable | Reusable |
Graduated | No |
Mouth Style | Narrow |
Sterility | Non-sterile |
Bottom Shape | Plain |
Closure Size | Stopper No. 8 |
Unit Size | Pack of 2 |
Capacity (English) | 64 oz. |
Description | Flask Erlenmeyer PP 2000mL |
Capacity (Metric) | 2000 mL |
Material | PPCO |
No. per Case | 6 |
No. per Pack | 2 |
Autoclavable | Autoclavable |
Disposable | Reusable |
Graduated | Yes |
Mouth Style | Narrow |
Sterility | Non-sterile |
Bottom Shape | Plain |
Closure Size | Stopper No. 10 |
Unit Size | Case of 6 |
Capacity (English) | 64 oz. |
Description | Flask Erlenmeyer PP 2000mL |
Capacity (Metric) | 2000 mL |
Material | PPCO |
No. per Case | 6 |
No. per Pack | 2 |
Autoclavable | Autoclavable |
Disposable | Reusable |
Graduated | Yes |
Mouth Style | Narrow |
Sterility | Non-sterile |
Bottom Shape | Plain |
Closure Size | Stopper No. 10 |
Unit Size | Pack of 2 |
Catalog Number | Specifications | Unit Size | Capacity (Metric) | Closure Size | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
4102-1000 | Case of 6 | 1000 mL | Stopper No. 8 | 239.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4102-0125 | Case of 24 | 125 mL | Stopper No. 3 | 178.00 Online price 339.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4102-0125PK also known as 4102-0125 | Pack of 12 | 125 mL | Stopper No. 3 | 89.00 Online price 200.60 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4102-0250 | Case of 12 | 250 mL | Stopper No. 6 | 214.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4102-0250PK also known as 4102-0250 | Pack of 6 | 250 mL | Stopper No. 6 | 126.20 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4102-1000PK also known as 4102-1000 | Pack of 2 | 1000 mL | Stopper No. 8 | 94.20 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4102-2000 | Case of 6 | 2000 mL | Stopper No. 10 | 298.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4102-2000PK also known as 4102-2000 | Pack of 2 | 2000 mL | Stopper No. 10 | 117.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|