Ethyl oleate, 98%, mixture of homologeous fatty acid esters, Thermo Scientific Chemicals
Ethyl oleate, 98%, mixture of homologeous fatty acid esters, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl oleate, 98%, mixture of homologeous fatty acid esters, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 410290010
Price (USD)/ Each
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS111-62-6
IUPAC Nameethyl (9E)-octadec-9-enoate
Molecular FormulaC20H38O2
InChI KeyLVGKNOAMLMIIKO-VAWYXSNFSA-N
SMILESCCCCCCCC\C=C\CCCCCCCC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Saponification value177 to 188 mg KOH/g
Appearance (Form)Clear viscous liquid
Acid value=<0.5 mg KOH/g
Density(20°C) 0.869 to 0.874
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only