Thermo Scientific Chemicals

Ethyl valerate, 99%, Thermo Scientific Chemicals

Catalog number: 410475000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl valerate, 99%, Thermo Scientific Chemicals

Catalog number: 410475000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 410475000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
539-82-2
IUPAC Name
ethyl pentanoate
Molecular Formula
C7H14O2
InChI Key
ICMAFTSLXCXHRK-UHFFFAOYSA-N
SMILES
CCCCC(=O)OCC
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Refractive index
1.3990 to 1.4010 (20°C, 589 nm)
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text