Thermo Scientific Chemicals

Ethylxanthic acid potassium salt, 97+%, Thermo Scientific Chemicals

Catalog number: 410501000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethylxanthic acid potassium salt, 97+%, Thermo Scientific Chemicals

Catalog number: 410501000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 410501000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
140-89-6
IUPAC Name
potassium (ethoxymethanethioyl)sulfanide
Molecular Formula
C3H5KOS2
InChI Key
JCBJVAJGLKENNC-UHFFFAOYSA-M
SMILES
[K+].CCOC([S-])=S
Appearance (Color)
Light yellow to yellow
Infrared spectrum
Conforms
Loss on drying
=<2 % (1 g, 105°C)
Appearance (Form)
Powder or crystalline powder or crystals
Titration with HClO4
97.0 to 103.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text