Thermo Scientific Chemicals

Fast Green FCF, pure, certified

Catalog number: 410530250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Fast Green FCF, pure, certified

Catalog number: 410530250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 410530250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2353-45-9
IUPAC Name
disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-hydroxy-2-sulfonatophenyl)methylium
Molecular Formula
C37H34N2Na2O10S3
InChI Key
XJBPDZVCYTYRIN-UHFFFAOYSA-L
SMILES
[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
Appearance (Form)
Powder
Absorption Ratio P-15/P+15
0.98 to 1.20
Infrared spectrum
Conforms
UV Lambda max
622 to 626 nm
Appearance (Color)
Bordeaux to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text