Thermo Scientific Chemicals

Fluorexon

Catalog number: 410630250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Fluorexon

Catalog number: 410630250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 410630250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1461-15-0
IUPAC Name
2-{[(7'-{[bis(carboxymethyl)amino]methyl}-3',6'-dihydroxy-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-2'-yl)methyl](carboxymethyl)amino}acetic acid
Molecular Formula
C30H26N2O13
InChI Key
DEGAKNSWVGKMLS-UHFFFAOYSA-N
SMILES
OC(=O)CN(CC(O)=O)CC1=CC2=C(OC3=CC(O)=C(CN(CC(O)=O)CC(O)=O)C=C3C22OC(=O)C3=CC=CC=C23)C=C1O
Suitability
(for Ca determination) Passes test
Infrared spectrum
Conforms
UV spectrum
Complies
Appearance (Color)
Yellow to orange-brown
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text