HT Supplement (50X)
Product Image

HT Supplement (50X)

Gibco™ HT Supplement is a liquid mixture of sodium hypoxanthine (5 mM) and thymidine (0.8 mM). HT-supplemented medium is suitableRead more
Have Questions?
Catalog NumberQuantity
41065012100 mL
Catalog number 41065012
Price (EUR)
28,93
Each
-
Add to cart
Quantity:
100 mL
Price (EUR)
28,93
Each
Add to cart
Gibco™ HT Supplement is a liquid mixture of sodium hypoxanthine (5 mM) and thymidine (0.8 mM). HT-supplemented medium is suitable for post-selection rescue to overcome the effects of residual intracellular aminopterin. Once the de novo biosynthesis pathway for nucleosides has been reestablished, HT supplementation can be discontinued.

cGMP Manufacturing and Quality System
Gibco™ HT Supplement is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ HT Supplement product made in our Grand Island facility (11067-030). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMammalian Cells
Concentration50 X
Quantity100 mL
Shelf Life18 Months
Shipping ConditionRoom Temperature
FormLiquid
Product TypeHT Supplement
SterilitySterile-filtered
pH9 to 11
Unit SizeEach
Contents & Storage
Storage conditions: 2-8°C
Shipping conditions: Ambient
Shelf life: 18 months from date of manufacture

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link