Thermo Scientific Chemicals

Fluosilicic acid, 25 wt.% aqueous solution, Thermo Scientific Chemicals

Catalog number: 410715000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Fluosilicic acid, 25 wt.% aqueous solution, Thermo Scientific Chemicals

Catalog number: 410715000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 410715000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16961-83-4, 7732-18-5
Appearance (Color)
Clear colorless
Assay
19 to 20.6 % (total fluorine)
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text