Thermo Scientific Chemicals

Gallic Acid 98%, Thermo Scientific Chemicals

Catalog number: 410862500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Gallic Acid 98%, Thermo Scientific Chemicals

Catalog number: 410862500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 410862500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
149-91-7
IUPAC Name
3,4,5-trihydroxybenzoic acid
Molecular Formula
C7H6O5
InChI Key
LNTHITQWFMADLM-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC(O)=C(O)C(O)=C1
Appearance (Color)
White to beige
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration with NaOH
97.5 to 102.5 %
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text