Geraniol, 99%
Geraniol, 99%
Geraniol, 99%
Thermo Scientific Chemicals

Geraniol, 99%

CAS: 106-24-1 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 410900050
Price (USD)
18.65
Special offer
Online exclusive
21.50
Save 2.85 (13%)
Each
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
18.65
Special offer
Online exclusive
21.50
Save 2.85 (13%)
Each
Add to cart
Chemical Identifiers
CAS106-24-1
IUPAC Name(2E)-3,7-dimethylocta-2,6-dien-1-ol
Molecular FormulaC10H18O
InChI KeyGLZPCOQZEFWAFX-JXMROGBWSA-N
SMILESCC(C)=CCC\C(C)=C\CO
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4750 to 1.4780 (20°C, 589 nm)
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
With the Thermo Scientific Protein:Protein Interaction (PPI) kits, what methods can be used to detect the prey protein?
How many reactions can be performed using one Thermo Scientific Protein:Protein Interaction (PPI) kit?
How is the tagged protein detected on the membrane using the Thermo Scientific Protein:Protein Interaction (PPI) kits?
Are all of the conditions for binding the prey protein to the bait protein described in the Thermo Scientific Protein:Protein Interaction (PPI) kits?
What types of Thermo Scientific Protein:Protein Interaction (PPI) kits are available?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0472789Certificate of AnalysisMay 21, 2025410901000, 410900025, 410905000, 410900250, 410900050
A0460157Certificate of AnalysisFeb 14, 2024410901000, 410900025, 41090036, 410905000, 41090028, 410900250, 410900050
2691490Certificate of AnalysisMay 09, 202341090036, 41090028, 410901000, 410905000, 410900050, 410900025, 410900250
2562478Certificate of AnalysisFeb 09, 202341090036, 41090101, 410901000, 410905000, 410900050, 410900025, 410900250
2511083Certificate of AnalysisFeb 09, 202341090036, 41090101, 410901000, 410905000, 410900050, 410900025, 410900250
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection