Thermo Scientific Chemicals

Geraniol, 99%, Thermo Scientific Chemicals

Catalog number: 410901000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Geraniol, 99%, Thermo Scientific Chemicals

Catalog number: 410901000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 410901000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-24-1
IUPAC Name
(2E)-3,7-dimethylocta-2,6-dien-1-ol
Molecular Formula
C10H18O
InChI Key
GLZPCOQZEFWAFX-JXMROGBWSA-N
SMILES
CC(C)=CCCC(C)=CCO
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=98.5 %
Refractive index
1.4750 to 1.4780 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text