Thermo Scientific Chemicals

Glycine hydrochloride, 98+%

Catalog number: 411015000
500 g, Glass Bottle, Each
Thermo Scientific Chemicals

Glycine hydrochloride, 98+%

Catalog number: 411015000
500 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 411015000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6000-43-7
IUPAC Name
hydrogen 2-aminoacetic acid chloride
Molecular Formula
C2H6ClNO2
InChI Key
IVLXQGJVBGMLRR-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NCC(O)=O
Melting point
175°C to 180°C
Water
=<1.0 % (K.F.)
Appearance (Color)
White to almost white
Infrared spectrum
Conforms
Anticaking agent
0.5 % (SiO2)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text