Thermo Scientific Chemicals

Glycolic acid 70% in water, Thermo Scientific Chemicals

Catalog number: 411030250
25 kg, Metal drum, Each
Thermo Scientific Chemicals

Glycolic acid 70% in water, Thermo Scientific Chemicals

Catalog number: 411030250
25 kg, Metal drum, Each
Quantity
Packaging
Catalog number: 411030250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79-14-1
IUPAC Name
2-hydroxyacetic acid
Molecular Formula
C2H4O3
InChI Key
AEMRFAOFKBGASW-UHFFFAOYSA-N
SMILES
OCC(O)=O
Appearance (Form)
Solution
Titration with NaOH
65 to 72 % w/w
Appearance (Color)
Clear colorless to yellow to light amber

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text