Thermo Scientific Chemicals

Glyoxylic Acid 50% in Water, Thermo Scientific Chemicals

Catalog number: 411060025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Glyoxylic Acid 50% in Water, Thermo Scientific Chemicals

Catalog number: 411060025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 411060025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
298-12-4
IUPAC Name
2-oxoacetic acid
Molecular Formula
C2H2O3
InChI Key
HHLFWLYXYJOTON-UHFFFAOYSA-N
SMILES
OC(=O)C=O
Appearance (Form)
Solution
Refractive index
1.4130 to 1.4200 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow
Titration with NaOH
>=50 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text