Guanidine thiocyanate, 99%, for biochemistry
This product ships via ground from US northeast.
Guanidine thiocyanate, 99%, for biochemistry
Thermo Scientific Chemicals

Guanidine thiocyanate, 99%, for biochemistry

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 411111000
Price (USD)/ Each
98.10
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
98.10
Add to cart
Guanidine thiocyanate, 99%, for biochemistry
Catalog number411111000
Price (USD)/ Each
98.10
-
Add to cart
Chemical Identifiers
CAS593-84-0
IUPAC Nameguanidine; sulfanylformonitrile
Molecular FormulaC2H6N4S
InChI KeyZJYYHGLJYGJLLN-UHFFFAOYSA-N
SMILESSC#N.NC(N)=N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Infrared spectrumConforms
Titration Argentometric>=98.5 %
UVat 280 nm A: =<0.4
Sulfated ash=<0.05 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Guanidine thiocyanate is a useful tool for RNA isolation and protein solubilization. It is involved in the quantification of mRNA from hepatocytes. It acts as a medium for blood smaple lystae in order to maintain the integrity of nucleic acid. It is a chaotropic agent, mainly involved in the extraction of DNA and RNA. It is utilized to deactivate the influenza virus.

Solubility
Soluble in water and ethanol

Notes
Light sensitive. Incompatible with acids and strong oxidizing agents.
RUO – Research Use Only